
Gastrin I human

Synonym(s):
Gastrin I, heptadecapeptide, HG-17, Little gastrin I, human, [18-Pyroglutamic acid]big gastrin I-(18-34)-peptide amide, human, [pGlu18]-Big gastrin I fragment 18-34 amide, Gastrin I, Human
Empirical Formula (Hill Notation):
C97H124N20O31S
CAS Number:
Molecular Weight:
2098.20
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing