Skip to Content
Merck
All Photos(1)

Key Documents

05-23-2301

Sigma-Aldrich

Gastrin I, Human

Synthetic, human gastrin I. Peptide hormone produced in the stomach. Responsible for stimulation of gastric acid secretion. Delays gastric emptying and increases blood flow in the gastric mucosa.

Synonym(s):

Gastrin I, Human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C97H124N20O31S
CAS Number:
Molecular Weight:
2098.20
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

0.1% NH4OH, pH 8.0: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C97H124N20O31S/c1-49(2)39-68(114-95(146)71(43-54-46-100-59-18-11-9-16-57(54)59)116-97(148)73-19-12-37-117(73)76(121)48-102-85(136)60-24-30-74(119)104-60)93(144)110-65(29-35-81(130)131)91(142)109-64(28-34-80(128)129)90(141)108-63(27-33-79(126)127)89(140)107-62(26-32-78(124)125)88(139)106-61(25-31-77(122)123)87(138)103-50(3)84(135)113-69(41-52-20-22-55(118)23-21-52)86(137)101-47-75(120)105-70(42-53-45-99-58-17-10-8-15-56(53)58)94(145)111-66(36-38-149-4)92(143)115-72(44-82(132)133)96(147)112-67(83(98)134)40-51-13-6-5-7-14-51/h5-11,13-18,20-23,45-46,49-50,60-73,99-100,118H,12,19,24-44,47-48H2,1-4H3,(H2,98,134)(H,101,137)(H,102,136)(H,103,138)(H,104,119)(H,105,120)(H,106,139)(H,107,140)(H,108,141)(H,109,142)(H,110,144)(H,111,145)(H,112,147)(H,113,135)(H,114,146)(H,115,143)(H,116,148)(H,122,123)(H,124,125)(H,126,127)(H,128,129)(H,130,131)(H,132,133)

InChI key

GKDWRERMBNGKCZ-UHFFFAOYSA-N

General description

Synthetic, human gastrin I. Peptide hormone produced in the stomach. Responsible for stimulation of gastric acid secretion. Delays gastric emptying and increases blood flow in the gastric mucosa.
Synthetic, human gastrin I. Peptide hormone produced in the stomach. Responsible for stimulation of gastric acid secretion. Delays gastric emptying; increases blood flow in the gastric mucosa.

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

Pyr-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH₂ (Pyr = L-pyroglutamyl)

Physical form

Sold on the basis of peptide content.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Walsh, J.H. 1977. Fed. Proc.36, 1948.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service