Skip to Content
Merck
All Photos(1)

Documents

820023

TSKgel® BioAssist® Phenyl (10 µm) HPLC Columns

L x I.D. 5 cm × 7.8 mm, phenyl phase

Synonym(s):

TSKgel® Hydrophobic Interaction Chromatography (HIC) Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® BioAssist® Phenyl HPLC Column, phase phenyl, L × I.D. 5 cm × 7.8 mm, 10 μm particle size, PEEK hardware

material

PEEK hardware

product line

TSKgel®

manufacturer/tradename

Tosoh 20023

parameter

≤4 M salt concentration
≤50% organic solvent
1.2 mL/min flow rate
10-50 °C temperature
20 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

phenyl phase

particle size

10 μm

pore size

1000 Å

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

The most hydrophobic among the three TSKgel HIC columns, TSKgel Phenyl-5PW is an excellent choice to screen for the selectivity, retention and recovery of most biomolecules. Featuring a 1000Å pore size in a polymethacrylate base material, the Phenyl-5PW column is stable in either acid or caustic cleaning regimens and provides excellent access to larger molecules with low diffusion coefficients. Stainless steel is the standard column format for TSKgel Phenyl-5PW. Glass and PEEK column housings are available for samples that may adsorb to stainless steel. The alternative column formats are also suitable for general applications.
Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity and allow selective binding and desorption of proteins. However, HIC operates at significantly lower binding energy and uses aqueous mobile phases. These characteristics provide a gentle technique that is less likely to disturb protein conformation. As a result, HIC generally provides better activity recovery.

TSKgel BioAssist Phenyl columns contain the same particles as Phenyl-5PW columns, but are packed in PEEK column hardware rather than stainless steel. BioAsist Phenyl (Phenyl-5PW), TSKgel Ether-5PW and Butyl-NPR resin-based columns provide a range of hydrophobicities for chromatographic optimization. Ammonium sulfate concentration can be minimized by using a more hydrophobic packing, such as BioAssist Phenyl. We recommend an Ether-5PW column for purifying very hydrophobic proteins. TSKgel BioAssist Phenyl and TSKgel Ether-5PW packings are based on TSKgel G5000PW resin - 10 μm particles with 1000 Å pores. TSKgel Butyl-NPR columns, which are less hydrophobic than Ether-5PW, is prepared from 2.5 μm nonporous particles, allowing for rapid analyses and semi-micropreparative applications. All three column types can be cleaned with 0.2 M NaOH. Scale-up can be performed by using corresponding Toyopearl bulk resins.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service