Key Documents
22144-U
Supelco
Gas Sampling Bulb
size 1000 mL, glass stopcock
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
Pricing and availability is not currently available.
material
glass stopcock
technique(s)
whole air sampling: suitable
size
1000 mL
application(s)
air monitoring
environmental
industrial hygiene
General description
Use these glass bulbs to trap and transport a gas sample, then remove an aliquot (through the plug-type septum) for analysis. Oven annealed to resist damage during transportation and on-site use. Available with PTFE or high vacuum glass stopcocks. One half-hole cylindrical Thermogreen septum is included with each bulb.
Choose from one of the most recent versions:
Certificates of Analysis (COA)