Documents
20439
Supelco
Plastic Column Cap
for use with 1/4 in. O.D. column
About This Item
UNSPSC Code:
41121800
packaging
pkg of 100 ea
technique(s)
gas chromatography (GC): suitable
fitting
female cap for 1/4 in. O.D. column (slip-on)
compatibility
for use with 1/4 in. O.D. column
General description
These plastic caps will easily slip over the ends of packed GC columns to protect them during storage.
Application
Plastic column cup maybe used to close the immunoaffinity chromatography column used for separating cross-linked protein complexes adhering to P1 protein of Mycoplasma pneumonia.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
G Layh-Schmitt et al.
Microbiology (Reading, England), 146 ( Pt 3), 741-747 (2000-04-04)
Adherence of Mycoplasma pneumoniae to host cells requires several mycoplasmal membrane proteins and cytoskeleton-like proteins in addition to the adhesin P1, a transmembrane protein of 170 kDa. To analyse interactions of the P1 adhesin with other membrane proteins or with