Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1055

Sigma-Aldrich

Anti-PGAM5 Antibody, clone 2J4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Bcl-XL-binding protein v68, Phosphoglycerate mutase family member 5, Serine/threonine-protein phosphatase PGAM5 mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J4, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 28.02 kDa
calculated mol wt 32 kDa
observed mol wt ~32 kDa
observed mol wt ~34 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PGAM5(192111)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2J4 is a ZooMAb rabbit recombinant monoclonal antibody that detects both isoforms of human Phosphoglycerate mutase family member 5 (PGAM5). It targets an epitope within 86 amino acids from the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 86 amino acids from the N-terminal half of human Phosphoglycerate mutase family member 5 (PGAM5).

Application

Anti-PGAM5, clone 2J4 ZooMAb, Cat. No. ZRB1055, is a Rabbit monoclonal antibody that specifically targets PGAM5 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PGAM5 in lysates from HEK293 cells, mouse testis, and human brain tissue.

Immunohistochemistry (Paraffin) Analysis: 1:100 dilution from a representative lot detected PGAM5 in human testis and human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant PGAM5 with a KD of 3.0 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PGAM5 in MCF7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Serine/threonine-protein phosphatase PGAM5, mitochondrial (UniProt: Q96HS1; also known as EC:3.1.3.16, Bcl-XL-binding protein v68, Phosphoglycerate mutase family member 5) is encoded by the pgam5 GENE (Gene ID: 192111) in human. PGAM5 is a single-pass mitochondrial membrane protein that displays phosphatase activity for serine/threonine residues and is shown to dephosphorylate and activate MAP3K5 kinase. It does not display any phosphoglycerate mutase activity. PGAM5 usually exists as a dimer, but can also form ternary complex with Nuclear Factor, Erythroid 2 Like 2 (NFE2L2) and Kelch Like ECH Associated Protein 1 (KEAP1). Its transmembrane domain is localized to amino acids 7 to 29. A 35 amino acid sequence at its N-terminal function as a non-cleaved mitochondrial targeting sequence that targets the protein to the cytosolic side of the outer mitochondrial membrane. Two isoforms of PGAM5 have been described that are produced by alternative splicing. Both isoforms are reported to be phosphorylated by RIPK1/RIPK3 complex under conditions of necrosis and this phosphorylation is shown to increase PGAM5 phosphatase activity. Overexpression of isoform 1 results in clustering of mitochondria around the nucleus whereas isoform 2 overexpression results in the formation of disconnected punctuate mitochondria distributed throughout the cytoplasm. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service