Skip to Content
Merck
All Photos(1)

Key Documents

Z744273

Wilmad® Suprasil® Low Temperature Aqueous Cell

volume 100 μL, pack of 1 ea

Synonym(s):

EPR aqueous cell, aqueous cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-808-Q

volume

100 μL

General description

Small Volume TE102 Variable Temperature Aqueous Cell works with most common Variable Temperature Dewar Inserts. This Aqueous Cell contains Suprasil in critical parts.

Application

EPR

Legal Information

Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service