Skip to Content
Merck
All Photos(1)

Key Documents

SRP0518

Sigma-Aldrich

Human PD-L1 (CD274), FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

B7 homolog 1, B7-H1, CD274, PD-L1, PDCD1L1, Programmed Cell Death 1 Ligand 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

54 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD274(29126)

General description

Human secreted Programmed Cell Death 1 Ligand 1 (PD-L1)-Fc fusion protein, also known as CD274 and B7 homolog 1 (B7- H1), GenBank Accession No. NM_014143, amino acids 19-239, fused at the Cterminus with the Fc portion of human IgG1, with C-terminal Avi-tag expressed in a HEK293 cell expression system and enzymatically biotin-labeled using Avi-tag technology. MW = 54 kDa.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service