Skip to Content
Merck
All Photos(1)

Key Documents

SRE0016

Sigma-Aldrich

MATP+

substrate for analysis of acetylcholinesterase (AChE) activity

Synonym(s):

1,1-dimethyl-4-acetylthiomethylpiperidinium iodide, acetylcholinesterase substrate, MATP+

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20INOS
Molecular Weight:
329.24
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

MATP+,

storage temp.

2-8°C

Quality Level

SMILES string

CC(SCC1CC[N+](C)(C)CC1)=O.[I-]

InChI

1S/C10H20NOS.HI/c1-9(12)13-8-10-4-6-11(2,3)7-5-10;/h10H,4-8H2,1-3H3;1H/q+1;/p-1

InChI key

WLNVGCIMLLIQAL-UHFFFAOYSA-M

General description

1,1-Dimethyl-4-acetylthiomethylpiperidinium iodide (MATP+) is a substrate for analysis of acetylcholinesterase (AChE) activity.  MATP+ is highly selective for specific detection of AChE activity, and thus is advantageous over other AChE substrates like acetylcholine, which can react with multiple enzymes like AChE and also butylcholinesterase (BChE).  Thus with MATP+ in the AChE assay, there is no need to include BChE inhibitors in the protocol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Kikuchi et al.
Biological & pharmaceutical bulletin, 33(4), 702-706 (2010-04-23)
The activity of acetylcholinesterase (AChE) is measured to obtain pathological information about the cholinergic system in various disease states and to assess the effect of AChE inhibitors. Using Ellman's method that is commonly used in such examinations, butyrylcholinesterase inhibitors must
Hachiro Sugimoto
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 124(3), 163-170 (2004-08-31)
Enhancement of the activity of cholinergic neurons has been regarded as one of the most promising methods for treating Alzheimer's disease (AD). Donepezil is a representative acetylcholinesterase inhibitor (AChEI) and is a great success among the AChEI drugs. AChEIs are
Nobuo Tsuruoka et al.
PloS one, 7(11), e50824-e50824 (2012-12-05)
Diketopiperazines (DKPs) are naturally-occurring cyclic dipeptides with a small structure and are found in many organisms and in large amounts in some foods and beverages. We found that a chicken essence beverage, which is popular among Southeast Asians as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service