Skip to Content
Merck
All Photos(1)

Key Documents

SAE0154

Sigma-Aldrich

E64 Ready Made solution

Non-freezing solution, 1 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

Quality Level

form

ready-to-use solution

shipped in

wet ice

storage temp.

−20°C

General description

E-64 is a cysteine protease inhibitor that effectively inhibits various cysteine proteases such as: cathepsin K, cathepsin L, cathepsin S known also as N-[N-(L-3-trans-carboxyoxiran-2-carbonyl)-L-leucyl]-agmatine, E-64 was isolated from the mold Aspergillus japonicus TPR-64.

Biochem/physiol Actions

E-64 is an highly selective, potent and irreversible cysteine protease inhibitor. The trans-epoxysuccinyl group of E-64 is its active moiety, and irreversibly binds to an active thiol group of many cysteine proteases, such as papain, actinidase, and cathepsins B, H and L, to form a thioether linkage.

Preparation Note

This E-64 Ready Made Solution is supplied as a proprietary 2.8 mM non-freezing solution formulation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service