Skip to Content
Merck
All Photos(2)

Key Documents

M9131

Sigma-Aldrich

2′-O-Monosuccinyladenosine 3′:5′-cyclic monophosphate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N5O7P
CAS Number:
Molecular Weight:
421.30
EC Number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥98%

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

(Nc1ncnc2c1ncn2C1OC2COP(=O)(O)OC2C1OC(=O)CCC(=O)O)

InChI

1S/C14H16N5O9P/c15-12-9-13(17-4-16-12)19(5-18-9)14-11(27-8(22)2-1-7(20)21)10-6(26-14)3-25-29(23,24)28-10/h4-6,10-11,14H,1-3H2,(H,20,21)(H,23,24)(H2,15,16,17)

InChI key

QNPSLGPIZRJDAN-UHFFFAOYSA-N

Related Categories

Application

2′-O-Monosuccinyladenosine 3′:5′-cyclic monophosphate is a cAMP analog that may be used with other cAMP analogs to evaluate cAMP mediate processes within cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinay Bulusu et al.
Biochimica et biophysica acta, 1794(4), 642-654 (2008-12-30)
Adenylosuccinate lyase (ASL) catalyzes two distinct but chemically similar reactions in purine biosynthesis. The first, exclusive to the de novo pathway involves the cleavage of 5-aminoimidazole-4-(N-succinylcarboxamide) ribonucleotide (SAICAR) to 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) and fumarate and the second common to both
S C Wen et al.
Endocrinology, 116(3), 935-944 (1985-03-01)
Adrenal cortical cells are known to export cAMP and have binding proteins and cAMP-dependent protein kinase activity associated with their plasma membranes. Because these properties suggest a function for extracellular cAMP, we have undertaken a search for specific cell surface
R A Richman et al.
Journal of cyclic nucleotide research, 6(6), 461-468 (1980-01-01)
Antisera to cyclic AMP and cyclic GMP were obtained by immunizing rabbits with antigens prepared by conjugating the 2'0-succinyl derivative of the cyclic nucleotides to thyroglobulin. The cyclic nucleotide-thyroglobulin conjugates were injected intradermally into multiple sites on the backs of
Michael Newton et al.
Analytical biochemistry, 402(2), 129-136 (2010-04-08)
Nitric oxide (NO) mediates intercellular signaling through activation of its receptor, soluble guanylyl cyclase (sGC), leading to elevation of intracellular guanosine 3',5'-cyclic monophosphate (cGMP) levels. Through this signal transduction pathway, NO regulates a diverse range of physiological effects, from vasodilatation
D Moreno-Delgado et al.
Neuroscience, 164(3), 1244-1251 (2009-09-09)
Histamine H(3) autoreceptors induce a negative feedback on histamine synthesis and release. While it is known that cAMP/cAMP dependent protein kinase (PKA) and Ca(2+)/CaMKII transduction pathways mediate H(3) effects on histamine synthesis, the pathways regulating neuronal histamine release are poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service