K2643
KDR (789-end), active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLK1, VEGFR, VEGFR2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
53-71 nmol/min·mg
mol wt
~110 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... KDR(3791)
Biochem/physiol Actions
KDR (kinase insert domain receptor) is a transmembrane tyrosine kinase receptor which interacts with VEGF (vascular endothelial growth factor) and helps in mediating VEGF downstream angiogenic effects. Blocking KDR suppresses tumor growth and abrogates tumor cell invasion.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Z Zhu et al.
Investigational new drugs, 17(3), 195-212 (2000-02-09)
Angiogenesis, the formation of new blood vessels, is essential for both tumor growth and metastasis. Recent advances in our understanding of the molecular mechanisms underlying the angiogenesis process and its regulation have led to the discovery of a variety of
Baohui Qi et al.
Archiv der Pharmazie, 346(8), 596-609 (2013-07-12)
Novel quinoline derivatives bearing acyclic semicarbazones were prepared and their chemical structures as well as the relative stereochemistry were confirmed. All the synthesized compounds were evaluated for their c-Met kinase inhibitory activity and their cytotoxicity against the cell lines HT-29
Mingze Qin et al.
Archiv der Pharmazie, 346(11), 840-850 (2013-10-12)
A series of 4-(2-fluorophenoxy)-2-(1H-tetrazol-1-yl)pyridines bearing semicarbazone moieties were synthesized and evaluated for their in vitro antitumor potency. Some of the compounds (10b, 10c, 10e-10h, 10m-10p, 10r, and 11b) exhibited moderate to excellent antitumor activity as compared to sorafenib and PAC-1
G Neufeld et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 13(1), 9-22 (1999-01-05)
Vascular endothelial growth factor (VEGF) is a highly specific mitogen for vascular endothelial cells. Five VEGF isoforms are generated as a result of alternative splicing from a single VEGF gene. These isoforms differ in their molecular mass and in biological
Chao Sun et al.
Scientific reports, 5, 9774-9774 (2015-05-30)
CHIP (c-terminal Hsp70-interacting protein) is an E3 ligase which may play different roles in different cancers. The elucidation of the VHL-HIF-1α (hypoxia inducible factor-1α)-VEGF (vascular endothelial growth factor) pathway has led to the development of targeted therapy in renal cell
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service