Key Documents
HS120834
Mega Rack® tube rack
white polypropylene
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
Pricing and availability is not currently available.
material
white polypropylene
feature
to hold 120 x 15 mL tubes or 20 mm round bottom tubes
manufacturer/tradename
Heathrow Scientific 120834
L × W × H
377 mm × 301 mm × 91 mm
General description
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage. Each rack has 120 wells and a well diameter of 20 mm.
Features and Benefits
  • Sturdy, durable material for easy and secure transport of clinical and research tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Reusable, easy to clean and Autoclavable
  • Place different test groupings into one rack eliminating the need for multiple racks
  • Stackable for staging and saving space
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)