Key Documents
HS120356
TracRack® Storage Rack
polypropylene, Holds 64 x 1.5-2 mL tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
Pricing and availability is not currently available.
material
polypropylene
feature
Holds 64 x 1.5-2 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120356
technique(s)
PCR: suitable
well diam.
12.3 mm
General description
Take what you need and store the rest. Removing and working with samples from cold storage has never been easier. Eliminate the hassle of taking an entire rack of samples out of cold storage with the tracrack that enables lab professionals to remove one row of samples at a time.
Eight individual segments are nested in a 2" polypropylene film box that allows users to easily open, identify and remove only the segments of samples needed.
Once the segments are removed, simply replace the box lid and leave the remaining samples in storage.
Each tube segment is made from durable polypropylene and allows for easy working while saving valuable counter space.
Two versions are available, tracrack that accomodates 1.5-2 mL cyro and micro centrifuge tubes, and the reversible tracrack micro which will hold 64 x 0.5 mL microtubes on one side and 96 x 0.2 mL micro tubes on the other.
Features and Benefits
  • segments made from polypropylene
  • eight segments per box
  • outer box is made from thin film polypropylene material
  • store down to temperatures of -80C
  • moisture resistant, significantly reducing the change of mold
  • removable segments allow outer shell to be used for storage of other small laboratory products
  • light tight box for light sensitive materials
  • 2" box fits in standard freezer racks
Other Notes
Wells have a center depression to stabilize conical tubes and a cryogenic vial lock for one handed cap removal.
Legal Information
TracRack is a registered trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)