Skip to Content
Merck
All Photos(2)

Key Documents

GBL246879

Grace Bio-Labs ProPlate® tray and cover

for stainless steel spring clip modules, holds 3 slide modules

Synonym(s):

slide holder, slide module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.15
external size:
85.3 mm × 127.5 mm
Pricing and availability is not currently available.

sterility

non-sterile

manufacturer/tradename

Grace Bio-Labs 246879

external size

85.3 mm × 127.5 mm

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Organize up to 3 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service