Key Documents
GBL103259
Grace Bio-Labs CultureWell removal tool
for removable chambered coverglass
Synonym(s):
chambered coverglass
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
acrylic
blue
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Grace Bio-Labs 103259
external size
53 mm × 100 mm
General description
Designed to separate the chambered upper structure from the coverglass and leave a removable gasket on the coverglass.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)