Key Documents
CLS3914
Corning® Medium Bind 1 x 8 Stripwell 96 well plates
96 well plate, 1 x 8 Stripwell 8-well strips, Medium Bind, polystyrene, flat bottom, black, non-sterile, 100/cs
Synonym(s):
1 x 8 strip plate, 8 well strip plate, 96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene plate
flat bottom black wells
polystyrene plate
size:
96 wells
material
black polystyrene plate
flat bottom black wells
polystyrene plate
sterility
non-sterile
feature
lid: no
skirt
plate format: 96 well Stripwell 8-well strips
packaging
case of 100
pack of 25
manufacturer/tradename
Corning 3914
size
96 wells
well volume
360 μL
well working volume
75-200 μL
wells
96 wells , stripwell
suitability
suitable for (in vitro diagnostic assays)
binding type
non-treated surface
General description
Corning® Medium Bind 1 x 8 Stripwell 96 Well Plates
  • Designed for in vitro diagnostic assays
  • Flat bottom strips are designed to easily break apart and are pre-assembled in an "egg-crate" style strip holder that allows each individual well to be positioned back into the plate once broken
  • Black wells have low background fluorescence, minimal light scatter and reduced crosstalk
  • White wells enhance luminescent signals, have low background luminescence and fluorescence, and prevent well-to-well crosstalk
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Recommended working volume of 75 to 200 μL
  • 1x8 Strips fit only one way into the strip holder, eliminating the chance of misorientation
Legal Information
Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)