Skip to Content
Merck
All Photos(1)

Key Documents

HF1

94101908, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HF1, 94101908

biological source

rat liver

packaging

tube of 5 μg 94101908-DNA-5UG
pkg of vial of cells 94101908-1VL

growth mode

Adherent

karyotype

Modal no. 100

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma Hybrid [H5xFao]

Cell Line Description

HF1 is a clone of hybrid cells resulting from the fusion of H5 and Fao. Expression of the majority of hepatic functions are extinguished. The cells contain a number of chromosomes corresponding to the sum of the parental numbers. HF1 variant which lacks the complete set of tissue-specific functions, appears to be a good candidate for regulatory mutants which control the orderly temporal expression of tissue specific functions during development.

Application

Analysis of gene expression by molecular techniques.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 9x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service