Skip to Content
Merck
All Photos(1)

Key Documents

Y0001277

Hydroxyethyl starch (medium MW)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydroxyethyl starch

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

starch

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O.OCCOC[C@H]2O[C@H](OCCO)[C@H](OCCO)[C@@H](OCCO)[C@@H]2OCCO

InChI

1S/C16H32O11.C6H12O6/c17-1-6-22-11-12-13(23-7-2-18)14(24-8-3-19)15(25-9-4-20)16(27-12)26-10-5-21;7-1-2-3(8)4(9)5(10)6(11)12-2/h12-21H,1-11H2;2-11H,1H2/t12-,13-,14+,15-,16+;2-,3-,4+,5-,6+/m11/s1

InChI key

DNZMDASEFMLYBU-RNBXVSKKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyethyl starch (medium MW) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

  1. C'EST QUOI LE MW DE CE PRODUIT?

    1 answer
    1. This is a product of the European Pharmacopoeia. The average molecular weight is reported on the product leaflet. The current lot reports an average Molecular weight of 130100 Da. Please see the link below to access the supplier leaflet:
      https://crs.edqm.eu/db/4DCGI/leaflet?leaflet=Y0001277_1.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service