SYNPLE-P082
Reagent-cartridge VHL-COOH-PEG2
Synple
Synonym(s):
Synple 2
Select a Size
¥72,100
Availability
Estimated to ship onFebruary 21, 2025
Select a Size
About This Item
¥72,100
Availability
Estimated to ship onFebruary 21, 2025
Recommended Products
1 of 4
This Item | SAB1300461 | SAB1300109 | SAB1300020 |
---|---|---|---|
IgG fraction of antiserum | IgG fraction of antiserum | IgG fraction of antiserum | IgG fraction of antiserum |
unconjugated | unconjugated | unconjugated | unconjugated |
dry ice | dry ice | dry ice | dry ice |
polyclonal | polyclonal | polyclonal | polyclonal |
buffered aqueous solution | buffered aqueous solution | buffered aqueous solution | buffered aqueous solution |
General description
Scale: up to 0.1 mmol
Linker: PEG2
E3 ligase target and ligand: von Hippel-Lindau (VHL), (S,R,S)-AHPC
Terminal functional group: carboxylic acid
Compatible substrates: amine
Cartridge contents:
- Supported carbodiimide and Oxyma Pure (coupling reagents)
- Supported carbonate (acid scavenger)
- SCX2 (for purification)
- Degrader building block linker reagent
- SYNPLE-P001 Reagent-cartridge CRBN-PEG1
- SYNPLE-P002 Reagent-cartridge CRBN-PEG2
- SYNPLE-P003 Reagent-cartridge CRBN-PEG3
- SYNPLE-P004 Reagent-cartridge CRBN-PEG4
- SYNPLE-P005 Reagent-cartridge CRBN-PEG5
- SYNPLE-P022 Reagent-cartridge VHL-PEG2
- SYNPLE-P023 Reagent-cartridge VHL-PEG3
- SYNPLE-P024 Reagent-cartridge VHL-PEG4
- SYNPLE-P025 Reagent-cartridge VHL-PEG5
- SYNPLE-P072 Reagent-cartridge CRBN-COOH-PEG2
- SYNPLE-P073 Reagent-cartridge CRBN-COOH-PEG3
- SYNPLE-P074 Reagent-cartridge CRBN-COOH-PEG4
- SYNPLE-P083 Reagent-cartridge VHL-COOH-PEG3
- SYNPLE-P084 Reagent-cartridge VHL-COOH-PEG4
Learn more about protein degradation.
Explore all our Protein Degrader Building Blocks.
Features and Benefits of Synple Platform
- Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
- Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety – all chemicals are contained in the system, minimizing the user’s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system – simple installation
- Reaction temperatures from 25 °C – 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Other Notes
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service