The specific surfactants present are not identified (ASTM standard MBAS analysis). The analyte values vary for each lot and not all lots contain all the analytes. Anionic Surfactant - WP, product QC114, contains sodium dodecyl sulfate.
QC1144
Anionic Surfactant - WP
certified reference material
About This Item
Recommended Products
grade
certified reference material
Quality Level
CofA
current certificate can be downloaded
packaging
ampule of 20 mL
manufacturer/tradename
RTC QCI-244
concentration
100% in H2O
technique(s)
UV/Vis spectroscopy: suitable
application(s)
environmental
format
matrix material
storage temp.
2-30°C
General description
Other Notes
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
I would like to know if the main ingredient is fluorine or other to check whether this product can be used as Capstone FS-66 Also, I would like to know how much concentration of Fluorosurfactant is and how much viscosity it has.
1 answer-
Helpful?
-
-
I'm getting Low results (3.1mg/l) for CRM QC1144 MBAS. We calibrate with SDS - could you confirm components of this CRM mixture as we may need to apply a factor to get the correct results if other surfactants (e.g. SDBS) are part of the mix? Thanks
1 answer-
All anionic surfactants present are calculated via the MBAS analysis method (methylene blue active anionic substances). The MBAS assay is an ASTM International standard technique for detecting anionic surfactants and this assay does not identify specific surfactants. For this product, the concentration values will vary from lot to lot. Some lots may not contain all of the analytes, however sodium dodecyl sulfate is present. This standard should be diluted according to instructions provided in the Certificate of Analysis - 10 mL of this standard is diluted in water to 1 liter for use. To discuss this further, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice and click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service