There are no recommended pathologies for this product. This product is intended to be used as a standard in chemistry laboratory methods, primarily compendial monographs, and is not intended for administration to animals or humans. This product may not be used for therapeutic or diagnostic purposes for animals or humans.
PHR1541
Leucovorin Calcium
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Folinic acid calcium salt, Calcium folinate
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to BP 923
traceable to Ph. Eur. C0250000
traceable to USP 1358004
API family
leucovorin
CofA
current certificate can be downloaded
packaging
ampule of 1 g
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Ca+2].N1(C(CNc3nc(nc(c31)[O-])N)CNc2ccc(cc2)C(=O)N[C@@H](CCC(=O)[O-])C(=O)O)C=O
InChI
1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1
InChI key
KVUAALJSMIVURS-ZEDZUCNESA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Per quale specifica patologie é consigliabile?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service