Skip to Content
Merck
All Photos(1)

Key Documents

PHL82325

Veratridine

phyproof® Reference Substance

Synonym(s):

3-Veratroylveracevine, Veracevine veratrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H51NO11
CAS Number:
Molecular Weight:
673.79
Beilstein:
78875
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

COc1ccc(cc1OC)C(=O)OC2CC[C@@]3(C)C4CCC5[C@]6(O)C[C@H](O)[C@@]7(O)C(CN8CC(C)CCC8[C@@]7(C)O)[C@]6(O)C[C@]35O[C@]24O

InChI

1S/C36H51NO11/c1-19-6-11-26-31(3,40)35(43)25(17-37(26)16-19)33(42)18-34-24(32(33,41)15-27(35)38)10-9-23-30(34,2)13-12-28(36(23,44)48-34)47-29(39)20-7-8-21(45-4)22(14-20)46-5/h7-8,14,19,23-28,38,40-44H,6,9-13,15-18H2,1-5H3/t19?,23?,24?,25?,26?,27-,28?,30-,31+,32+,33+,34+,35-,36-/m0/s1

InChI key

FVECELJHCSPHKY-FCSYLLAHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Opens voltage-dependent Na+ channels and prevents their inactivation.
Opens voltage-dependent Na+ channels and prevents their inactivation. This, in turn, opens voltage-activated calcium channels, thus increasing intracellular calcium content and inducing neurotransmitter release. Alkaloid neurotoxin which depolarizes excitable tissue; used to increase membrane sodium permeability. Veratridine is cytotoxic to chromaffin cells in vitro.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service