Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
technique(s)
X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
SRM 1297_cert
SRM 1297_SDS
Application
Other Notes
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdinum (Mo), Cobalt (Co)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Antimony (Sb), Arsenic (As), Bismuth (Bi), Boron (B), Lead (Pb), Niobium (Nb), Selenium (Se), Tantalum (Ta), Tellurium (Te), Tin (Sn), Titanium (Ti), Tungsten (W)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service