Skip to Content
Merck
All Photos(1)

Key Documents

BCR142R

Light sandy soil (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Analysis Note

For more information please see:
BCR142R

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Suska-Malawska et al.
Environmental monitoring and assessment, 191(2), 57-57 (2019-01-11)
In arid and semi-arid zones, atmospheric dust of different origins influences soil chemistry and plant biomass composition. Thus, studies on plant accumulation of heavy metals and rare earth elements (RREs) should include some assessments of potential eolian deposition. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service