Skip to Content
Merck
All Photos(1)

Key Documents

WP6110060

Millipore

Chemical Duty Pump, 100 V/50–60 Hz

Synonym(s):

Chemical Transfer Pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24
Pricing and availability is not currently available.

AC/DC input

100 V, 50–60 Hz

shipped in

ambient

Principle

Vacuum

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakurako Tanida et al.
Physical review. E, 101(3-1), 032607-032607 (2020-04-16)
Emergence and collapse of coherent motions of self-propelled particles are affected more by particle motions and interactions than by their material or biological details. In the reconstructed systems of biofilaments and molecular motors, several types of collective motion including a

Questions

  1. Is product WP6110060 CE Approved?

    1 answer
    1. Yes, the Millex High Output and Chemical Duty pumps are CE approved.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service