FCMAB215P
Milli-Mark® Anti-CD138-PE Antibody, clone MI15
clone MI15, Milli-Mark®, from mouse
Synonym(s):
Syndecan-1, SYND1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44
Recommended Products
biological source
mouse
Quality Level
conjugate
PE
antibody form
purified antibody
antibody product type
primary antibodies
clone
MI15, monoclonal
species reactivity
human
manufacturer/tradename
Milli-Mark®
technique(s)
flow cytometry: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SDC1(6382)
Related Categories
General description
CD138 is a transmembrane proteoglycan, bearing heparin sulphates and sometimes chondriotin sulfates. It consists of a 30.5 kDa core protein with 5 putative glycosaminoglycans attachment sites. CD138 is a member of the syndecan family. The reported Mr of CD138 ranges from 85,000 to 92,000, presumably depending on the glycosaminoglycan pattern of the molecule (Wijdenes, 1997). Within the hematopoietic system, CD138 is mainly confined to late stages of B-cell differentiation. It is typically expressed at high cellular density in plasma cells (Gattei, 1999). In mature tissues, CD138 is expressed by simple and stratified epithelia, fibroblasts, stratified keratinocytes and endothelial cells (Wijdenes, 1997). Anti-CD138, MI15, was included in the Sixth International Workshop and Conference on Human Leucocytes Differentiation Antigens, and studies by a number of different laboratories confirmed its reactivity with CD138 (Wijdenes, 1997).
Specificity
Antibody recognizes Human CD138 antigen
Application
Detect CD138 using this Milli-Mark Anti-CD138-PE Antibody, clone MI15 validated for use in FC.
Quality
Evaluated by flow cytometry using U266 cells
Target description
85-92 kDa (Wijdenes, 1997)
Legal Information
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service