Skip to Content
Merck
All Photos(1)

Key Documents

8.22034

Sigma-Aldrich

4-(Dimethylamino)-cinnamaldehyde

for synthesis

Synonym(s):

4-(Dimethylamino)-cinnamaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-[(CH3)2N]C6H4CH=CHCHO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352114
EC Index Number:
228-267-0
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

powder

mp

135-138 °C

storage temp.

2-30°C

SMILES string

N(C)(C)c1ccc(cc1)\C=C\C=O

InChI

1S/C11H13NO/c1-12(2)11-7-5-10(6-8-11)4-3-9-13/h3-9H,1-2H3/b4-3+

InChI key

RUKJCCIJLIMGEP-ONEGZZNKSA-N

Application

<ul>
<li><strong>High-resolution imaging of plant tissues:</strong> 4-dimethylaminocinnamaldehyde (DMACA) is used as a PA-specific fluorescent dye that allows localization of PAs at high resolution in cell walls and inside cells using confocal microscopy, revealing features of previously unreported wall-bound PAs (Chowdhury et al., 2022).</li>
<li><strong>Chromogenic reagent for dietary supplements:</strong> DMACA serves as a chromogenic reagent in a microplate spectrophotometric method, used for the analysis of indole-3-carbinol in dietary supplements, demonstrating its utility in enhancing the accuracy of supplement analysis (Chaisiwamongkhol et al., 2022).</li>
<li><strong>Development of fluorescent chemosensors:</strong> Utilized in the synthesis of the chemosensor NBF, which is employed for the detection of cyanide ions (Gao et al., 2022).</li>
</ul>

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 137 °C
Melting range (upper value): ≤ 140 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service