8.06455
Sodium formaldehydesulfoxylate hydrate
(stabilised with sodium carbonate) for synthesis
Synonym(s):
Sodium formaldehydesulfoxylate hydrate
About This Item
vapor pressure
2.68 hPa ( 20 °C)
Quality Level
form
powder
potency
6400 mg/kg LD50, oral (Rat)
pH
9.5-10.5 (20 °C, 100 g/L in H2O)
mp
63 °C
solubility
600 g/L
density
1.8 g/cm3 at 20 °C
bulk density
950 kg/m3
storage temp.
2-30°C
SMILES string
[Na+].[S](=O)(O)C[O-].O.O
InChI
1S/CH3O3S.Na.2H2O/c2-1-5(3)4;;;/h1H2,(H,3,4);;2*1H2/q-1;+1;;
InChI key
VJWVWUHJLZEHOU-UHFFFAOYSA-N
Application
- Latex Hydraulic Cement Additives: Discusses the role of Sodium formaldehyde sulfoxylate in modifying the initial hydration process of cement (D Gerry, TRANSPORTATION RESEARCH RECORD).
- Fabrication of Interconnected Plasmonic Spherical Silver Nanoparticles: Covers the utilization of Sodium formaldehyde sulfoxylate in forming Ag nanoparticles with enhanced LSPR peaks (S B. Aziz et al., Nanomaterials, 2019).
- Study on Synthesis and Properties of Vinyl Polyoxyethylene Ether Type Polycarboxylate Superplasticizer: Details the synthesis process involving Sodium formaldehyde sulfoxylate hydrate as a component (Z Jiang et al., IOP Conference Series, 2020).
- Copper Nanoparticles: Synthesis Methods and Its Light Harvesting Performance: Investigates the reaction conditions using Sodium formaldehyde sulfoxylate for copper nanoparticle synthesis (F Parveen et al., Solar Energy Materials and Solar Cells, 2016).
Analysis Note
Iron (Fe): ≤ 0.003 %
Identity (IR): passes test
This material contains approx. 5% sulfit resulting from the production process.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Muta. 2 - Repr. 2
Supplementary Hazards
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service