Skip to Content
Merck
All Photos(1)

Key Documents

5.34358

Sigma-Aldrich

Wnt-β-Catenin Signaling Inhibitor, FzM1

Synonym(s):

Wnt-β-Catenin Signaling Inhibitor, FzM1, Frizzled4 Modulator, FzM1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16N2O2S
CAS Number:
Molecular Weight:
360.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

SMILES string

C1=CC=C2C=C(C=CC2=C1)NC(=O)NC3=CC(=CC(=C3)C4=CC=CS4)O

InChI

1S/C21H16N2O2S/c24-19-12-16(20-6-3-9-26-20)11-18(13-19)23-21(25)22-17-8-7-14-4-1-2-5-15(14)10-17/h1-13,24H,(H2,22,23,25)

InChI key

OBTSACXGWYGCPK-UHFFFAOYSA-N

General description

A cell-permeable, biarylurea derivative that acts as a reversible inhibitor of Wnt-b-catenin signaling (EC50 = 5.74 µM). Acts by blocking the phosphorylation and nuclear translocation of beta-catenin thereby reduces the transcription of TCF/LEF-regulatory genes. Also shown to block TCF/LEF activation induced by lithium chloride. However, it does not affect GSK3-β or the APC destruction complex. Induces con-formational changes in Fz4-WT that ultimately allow it to inhibit β-catenin nuclear transport. Shown to promote HA-Fz4-FEVR plasma membrane localization in U87MG glioblastoma cells and Caco-2 cells (~ 10 µM). Also reported to abolish Norrin-dependent TCF/LEF activation and gene transcription in HEK293 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Frizzled 4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Generoso, S., et al. 2015. Nat. Chem. Bio.11, 280.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service