Skip to Content
Merck
All Photos(1)

Key Documents

521000-M

Millipore

Phenylarsine Oxide

A membrane-permeable protein tyrosine phosphatase inhibitor (IC₅₀ = 18 µM).

Synonym(s):

Phenylarsine Oxide, PAO, Oxophenylarsine, PTP Inhibitor IX, PDI Inhibitor II, Protein Disulfide Isomerase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5AsO
CAS Number:
Molecular Weight:
168.02
MDL number:
UNSPSC Code:
12352119

Quality Level

Assay

≥96% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: 100 mg/mL
DMSO: 50 mg/mL

storage temp.

10-30°C

SMILES string

[As](=O)c1ccccc1

InChI

1S/C6H5AsO/c8-7-6-4-2-1-3-5-6/h1-5H

InChI key

BQVCCPGCDUSGOE-UHFFFAOYSA-N

General description

A membrane-permeable phosphotyrosine phosphatase inhibitor (IC50 = 18 µM). Stimulates 2-deoxyglucose transport in insulin-resistant human skeletal muscle and activates P56lck protein tyrosine kinase. Blocks TNFα-dependent activation of the NF-κB in human myeloid ML-1a cells. PAO inhibits the protease activities of recombinant human caspases, as well as endogenous chicken caspases that are active in extracts of preapoptotic chicken DU249 cells (S/M extracts). At 100 µM, it also blocks the morphological changes of nuclear apoptosis in vitro and internucleosomal DNA fragmentation in S/M extracts without interfering with PARP or lamin A cleavage. Also known to cause complete and reversible inhibition of NADPH oxidase in human neutrophils.
A membrane-permeable protein tyrosine phosphatase inhibitor (IC50 = 18 µM). Stimulates 2-deoxyglucose transport in insulin-resistant human skeletal muscle and activates p56lck protein tyrosine kinase. Blocks TNF-α-dependent activation of NF-κB in human myeloid ML-1a cells. PAO inhibits the protease activities of recombinant human caspases as well as endogenous caspases that are active in extracts of pre-apoptotic chicken DU249 cells (S/M extracts).

Biochem/physiol Actions

Primary Target
Protein tyrosine phosphatase
Target IC50: 18 µM against protein tyrosine phosphatase

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Takahashi, A., et al. 1997. Exp. Cell Res.231, 123.
Carey, J.O., et al. 1995. Diabetes44, 1995.
Le Cabec, V., and Maridonneau-Parini, I. 1995. J. Biol. Chem.270, 2067.
Oetken, C., et al. 1995. Mol. Immunol.31, 1295.
Singh, S., and Aggarwal, B.B. 1995. J. Biol. Chem.270, 10631.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service