344826
α1,6-Fucosidase, Chryseobacterium meningosepticum, Recombinant, E. coli
This enzyme from Chryseobacterium (formerly Flavobacterium) meningosepticum catalyzes the hydrolysis of branched non-reducing terminal α1,6-linked fucose residues from the N-acetylglucosamine core of N-Linked oligosaccharides.
Synonym(s):
α1,6-Fucosidase, Chryseobacterium meningosepticum, Recombinant, E. coli, α-L-Fucoside Fucohydrolase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Recommended Products
recombinant
expressed in E. coli
Quality Level
form
liquid
specific activity
≥1.0 units/mL
≥1.5 units/mg protein
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
foreign activity
Proteases, none detected
shipped in
wet ice
storage temp.
2-8°C
General description
Note: 1 mU = 1 milliunit.
Recombinant, Chryseobacterium meningosepticum α1,6-fucosidase expressed in E. coli. This enzyme from Chryseobacterium (formerly Flavobacterium) meningosepticum catalyzes the hydrolysis of branched non-reducing terminal α1,6-linked fucose residues from the N-acetylglucosamine core of N-Linked oligosaccharides. The reducing terminus of the oligosaccharide must be covalently bonded to a reporter molecule, such as aminonaphthalene disulfonic acid (ANDSA), aminonaphthalene trisulfonate (ANTS), and 2-aminoanthanilic acid (2-AA). The one exception is that a terminal unbranched α1,3-fucose is cleaved in the absence of any reporter molecule. Does not cleave fucose from glycoproteins.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will release 1.0 µmol methylumbelliferone from 4-methylumbelliferyl-α-L-fucoside per min at 37°C, pH 5.0.
Physical form
In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.
Other Notes
Zheng, Q., et al. 2002. J. Biol. Chem.277, 39823.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service