Skip to Content
Merck
All Photos(1)

Key Documents

1.70234

Supelco

Zirconium standard solution

traceable to SRM from NIST ZrCl₄ in HCl 2 mol/l 1000 mg/l Zr Certipur®

Synonym(s):

Zirconium Calibration Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

product line

Certipur®

concentration

1000 mg/L Zr in 2 M HCl

technique(s)

AAS: suitable

pH

0.5 (20 °C in H2O)

density

1.034 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Zirconium standard solution may be used as a calibration standard in the determination of trace Zr(IV) in industrial Al-Li alloys by atomic absorption spectroscopy.[1]

Analysis Note

1000 mg/L Zr in 2 mol/L HCl, prepared with high-purityZrCl4, HCl, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new kinetic method for photometric determination of trace Zr (IV) in industrial Al-Li alloys
Pecev T, et al.
Analytical Chemistry, 52, 511-511 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service