Skip to Content
Merck
All Photos(1)

Key Documents

1.50082

Supelco

MANU-CART HPLC Coupling Kit

for LiChroCART 25-4 HPLC- cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

manu-CART Coupling kit for LiChroCART 25-4 HPLC- cartridges,

technique(s)

HPLC: suitable

Quality Level

storage temp.

no temp limit

Analysis Note

visual control: passes test

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service