Skip to Content
Merck
All Photos(1)

Key Documents

1.12254

Sigma-Aldrich

Molybdenum powder

99+

Synonym(s):

Molybdenum powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
EC Index Number:
231-107-2
NACRES:
NA.22

Quality Level

mp

2620 °C

density

10.2 g/cm3 at 20 °C

bulk density

1400 kg/m3

storage temp.

no temp limit

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Analysis Note

Assay (gravimetrically): ≥ 99.0 %
Identity (ICP-OES): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service