H8015 and 108166 are standards for testing wavelength accuracy, not for absorbance calibration.
1.08166
UV-VIS Standard 6: holmium oxide solution
according to Ph. Eur.
Synonym(s):
Holmium Oxide UV-VIS wavelength calibration sol. for spectrophotometer
About This Item
Recommended Products
grade
analytical standard
Quality Level
Agency
according to Ph. Eur.
technique(s)
UV/Vis spectroscopy: suitable
application(s)
environmental
industrial qc
format
mixture
storage temp.
15-25°C
Application
Features and Benefits
- It is a 40 g/L solution of holmium oxide in a solution of perchloric acid containing 141 g/L of HClO4
- Conveniently packed in 10 mL glass ampules. Each box includes 3 single-use ampules
- Stable solutions in glass ampoules
- Accreditation according to ISO 17025
- Traceability to standard reference material from the National Institute of Standards and Technology (NIST) and Physical-Technical Institute (PTB)
- Includes a certificate of analysis (CoA)
- Reliable and reproducible results
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT RE 2
Target Organs
Thyroid
Storage Class Code
5.1B - Oxidizing hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can the UV-VIS Standard 6 and Holmium wavelength calibration standard be used to test wavelength accuracy without absorbance values?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service