Skip to Content
Merck
All Photos(1)

Key Documents

1.03855

Millipore

GranuCult® prime SIMMONS Citrate Agar

according to ISO (10273 and FDA-BAM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

according to ISO (10273 and FDA-BAM)

Quality Level

form

powder

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

Escherichia coli
coliforms
nonselective for

General description

GranuCult® prime SIMMONS Citrate Agar is a nutrient rich synthetic medium which is designed for the differentiation of Gram-negative bacteria, primary among the Enterobacteriaceae, from food and animal feed, water and other materials, based on citrate utilization.
This culture medium is used for the ability of an organism to utilize citrate as the sole source of carbon and inorganic ammonium salt as sole source of nitrogen for metabolism with resultant alkalinity. The alkaline reaction is evidenced by a change in the colour of the pH indicator bromothymol blue from green (neutral pH) to blue (alkaline pH).
Simmons citrate agar is a synthetic modification of Koser′s medium with the addition of 1.5% agar and the pH indicator bromothymol blue.
Sodium citrate is the sole source of carbon and ammonium dihydrogen phosphate is the sole source of nitrogen. Magnesium sulfate is a cofactor for various metabolic reactions and sodium chloride maintains the osmotic balance. Bromothymol blue is the pH indicator and agar the solidifying agent.

Application

GranuCult® prime SIMMONS Citrate Agar is used for the differentiation of Gram-negative bacteria, primary among the Enterobacteriaceae, from food and animal feed, water and other materials, based on citrate utilization.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service