Skip to Content
Merck
All Photos(2)

Key Documents

857458M

Avanti

16:0 aldehyde

Avanti Research - A Croda Brand 857458M, methylene chloride solution

Synonym(s):

hexadecanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32O
CAS Number:
Molecular Weight:
240.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>90% (contains dimer, trimer, tetramer, TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (857458M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857458M

concentration

1 mg/mL (857458M-1mg)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC([H])=O

InChI

1S/C16H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h16H,2-15H2,1H3

InChI key

NIOYUNMRJMEDGI-UHFFFAOYSA-N

Application

16:0 aldehyde has been used as a component of aqueous buffer to study its enzyme activity on Hfd1p, aldehyde dehydrogenase (ALDH3A1) and ALDH3A2 enzymology.

Packaging

5 mL Clear Glass Sealed Ampule (857458M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan A Stefely et al.
Nature biotechnology, 34(11), 1191-1197 (2016-11-01)
Mitochondrial dysfunction is associated with many human diseases, including cancer and neurodegeneration, that are often linked to proteins and pathways that are not well-characterized. To begin defining the functions of such poorly characterized proteins, we used mass spectrometry to map
Mitochondrial protein functions elucidated by multi-omic mass spectrometry profiling
Stefely JA, et al.
Nature Biotechnology, 34(11), 1191-1191 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service