Key Documents
Z740242
QianCap gas purging set for Ace pressure tubes and bottles
QianCap-25-v2, fits no. 25 internal thread
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Features and Benefits
  • Two optional gas feed lines are available.
  • Maximum pressure: 60 psi
  • Provides optional high-pressure adapter (120 psi).
  • Septum for the system is disposable and made of blue PTFE face and white silicone backing.
  • Septa fits on Q-Tube-12 mL.
  • The blue PTFE liner provides unique chemical resistance and silicon backing has the resealing capability.
Other Notes
Pressure flasks/bottles not included
Legal Information
QianCap is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)