Z517550
Aldrich® round-bottom flask with septum-inlet
capacity 250 mL, joint: ST/NS 29/32
Synonym(s):
bottom flask, flask round, round flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
round bottom flask
two-neck
Joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
Related Categories
General description
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)