Key Documents
Z156566
Sigma-Aldrich
Aldrich® mineral-oil bubbler
threaded vacuum stopcock
Synonym(s):
bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
General description
With threaded vacuum stopcock and serrated inlet/outlet tubes. For controlling and monitoring gas flow at low (<2 psi) pressures.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Will this "stopcock" on Z156566 hold a vacuum worthy seal?
    1 answer
    1. The stopcock should hold a vacuum.
      Helpful?