Skip to Content
Merck
All Photos(1)

Key Documents

W387619

Sigma-Aldrich

S-Methyl thioacetate

natural, ≥96%, FG

Synonym(s):

1-Methyl sulfanyl ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6OS
CAS Number:
Molecular Weight:
90.14
FEMA Number:
3876
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.149
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥96%

refractive index

n/D 1.464

density

1.024

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cabbage; dairy; vegetable; sulfurous; cheesy

SMILES string

CSC(C)=O

InChI

1S/C3H6OS/c1-3(4)5-2/h1-2H3

InChI key

OATSQCXMYKYFQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service