GF36701287
Bismuth-lead alloy, Bi55.5Pb44.5
200g lump, 100mm max. lump size
Synonym(s):
BI076100
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12141706
NACRES:
NA.23
Recommended Products
form
lumps
manufacturer/tradename
Goodfellow 367-012-87
weight
200 g
max. lump size
100 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service