Skip to Content
Merck
All Photos(1)

Key Documents

BR781971

BRAND® microplate BRANDplates®, cellGrade

size 96 wells, size 330 μL, flat bottom, black, with transparent bottom

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
black wells
flat bottom
polystyrene
size:
330 μL
96 wells
working volume:
330 μL
sterility:
sterile
feature:
lid
skirt (F-bottom)
Pricing and availability is not currently available.

material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packed)

manufacturer/tradename

BRAND 781971

size

330 μL
96 wells

working volume

330 μL

color

black, with transparent bottom

Looking for similar products? Visit Product Comparison Guide

General description

BRANDplates® microplate, a 96-well plate for cell culture of adherent cell lines.

Features and Benefits

  • PS-surface with freely accessible chemical groups (carboxyl and hydroxyl), allowing easy binding of serum components and facilitating indirect cell adhesion
  • Hydrophilic surface compared to untreated PS, promoting cell attachment
  • Distinguishable with an orange color code and embossed alphanumeric coding for 96-well standard plates
  • Free from endotoxins, DNase, DNA, RNase, and non-cytotoxic substances
  • Made of sterile PS material and includes lids for aseptic cell culture conditions
  • BIO-CERT® CELL CULTURE QUALITY certified, ensuring the absence of human DNA, RNase, DNase, pyrogens, and non-cytotoxicity

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service