Skip to Content
Merck
All Photos(1)

Key Documents

931187

Sigma-Aldrich

Straight-Tip PVDF Tweezer with Carbon Tip

Synonym(s):

Gripper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
MA.24

Looking for similar products? Visit Product Comparison Guide

Application

Carbon tip tweezers help to avoid chipping the silicon wafer.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service