Skip to Content
Merck
All Photos(3)

Key Documents

905925

Sigma-Aldrich

[Pd(terpy)(2-Cl-phen)](BF4)2

≥95%

Synonym(s):

Ritter fluorination catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18B2ClF8N5Pd
CAS Number:
Molecular Weight:
727.95
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

>300 °C

storage temp.

2-8°C

SMILES string

ClC(C=C1)=NC2=C1C=CC3=CC=C[N]([Pd]45[N]6=C(C7=CC=CC=[N]75)C=CC=C6C8=CC=CC=[N]84)=C32

Application

[Pd(terpy)(2-Cl-phen)](BF4)2 was developed in the Ritter lab for the elecrophilic fluorination of arenes.
Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

This product is manufactured pursuant to a license with Studiengesellschaft Kohle mbH (US Patent Pending 16/083,160 and PCT/US2017/021563).

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kumiko Yamamoto et al.
Nature, 554(7693), 511-514 (2018-02-23)
Aryl fluorides are widely used in the pharmaceutical and agrochemical industries, and recent advances have enabled their synthesis through the conversion of various functional groups. However, there is a lack of general methods for direct aromatic carbon-hydrogen (C-H) fluorination. Conventional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service