Skip to Content
Merck
All Photos(1)

Key Documents

901610

Sigma-Aldrich

Bromoacetic acid solution

1.0 M in DMF

Synonym(s):

2-Bromoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2COOH
CAS Number:
Molecular Weight:
138.95
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

form

liquid

concentration

1.0 M in DMF

refractive index

n/D 1.4366

density

1.0264 g/mL

functional group

bromo
carboxylic acid

SMILES string

BrCC(O)=O

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

KDPAWGWELVVRCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Valuable building block offered as a solution in dimethylformamide for more convenient handling.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

85.0 °F

Flash Point(C)

29.44 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service