Skip to Content
Merck
All Photos(1)

Key Documents

901412

Sigma-Aldrich

Palmitic acid-7,7,8,8-d4

endotoxin tested, ≥98 atom % D, ≥99% (CP)

Synonym(s):

Hexadecanoic-7,7,8,8-d4 acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D4H28O2
CAS Number:
Molecular Weight:
260.45
MDL number:
UNSPSC Code:
12352106

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

solid

suitability

endotoxin tested

mass shift

M+4

SMILES string

CCCCCCCCC([2H])([2H])C([2H])([2H])CCCCCC(O)=O

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.


Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service