Skip to Content
Merck
All Photos(2)

Key Documents

775274

Sigma-Aldrich

Grubbs Catalyst® M201

Umicore

Synonym(s):

Dichloro[1,3-bis(2,6-diisopropylphenyl)-2-imidazolidinylidene](3-phenyl-1H-inden-1-ylidene)(triphenylphosphine)ruthenium(II), Grubbs Catalyst® M20 SIPr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H63Cl2N2PRu
CAS Number:
Molecular Weight:
1015.11
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

125-132 °C

storage temp.

2-8°C

SMILES string

Cl[Ru](=C1C=C(C2=CC=CC=C2)C3=C1C=CC=C3)(Cl)(C(C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6)=C7N(C8=C(C(C)C)C=CC=C8C(C)C)CCN7C9=C(C(C)C)C=CC=C9C(C)C

InChI

1S/C27H38N2.C19H15.C15H10.2ClH.Ru/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-6-12(7-3-1)15-11-10-13-8-4-5-9-14(13)15;;;/h9-14,18-21H,15-16H2,1-8H3;1-15H;1-9,11H;2*1H;/q;;;;;+2/p-2

InChI key

VQMXGRVEQOCJPP-UHFFFAOYSA-L

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service