Skip to Content
Merck
All Photos(1)

Key Documents

640379

Sigma-Aldrich

2-Aminopyridine-5-boronic acid pinacol ester

97%

Synonym(s):

(2-Aminopyridin-5-yl)boronic acid pinacol ester, 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-Pyridinamine, 6-Aminopyridine-3-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17BN2O2
CAS Number:
Molecular Weight:
220.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

131-135 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(N)nc2

InChI

1S/C11H17BN2O2/c1-10(2)11(3,4)16-12(15-10)8-5-6-9(13)14-7-8/h5-7H,1-4H3,(H2,13,14)

InChI key

YFTAUNOLAHRUIE-UHFFFAOYSA-N

Application

Substrate used in a microwave-assisted, four-component coupling process leading to amino substituted imidazopyridines.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin F Dimauro et al.
The Journal of organic chemistry, 72(3), 1013-1016 (2007-01-27)
The rapid and efficient synthesis of various 2,6-disubstituted-3-amino-imidazopyridines using a microwave-assisted one-pot cyclization/Suzuki coupling approach is described. The utility of a 2-aminopyridine-5-boronic acid pinacol ester as a robust and versatile building block for the synthesis of diverse compound libraries is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service